Learn More
Chemglass Life Sciences Cap, GL-45, White, SURE-LINK PTFE/Silicone Septum
SDP
Catalog No. 501311277
Encompass_Preferred
White polypropylene cap has a bonded septum using the Chemglass SURE-LINK process.This inert process bonds the PTFE/Silicone septum directly to the cap without adhesives.Autoclavable at 121 degreesC.
Catalog No. 50-131-1277 Supplier Chemglass Life Sciences Supplier No. CLS148310
White polypropylene cap has a bonded septum using the Chemglass SURE-LINK process.This inert process bonds the PTFE/Silicone septum directly to the cap without adhesives.Autoclavable at 121 degreesC.