Learn More
Med Vet International Syringe Caps, Polypropylene, Non-Sterile, Fits all luer slip and luer lock syringes. White, 100/Bag
Catalog No. 502522431
Encompass_Preferred
These caps used for sealing syringes used in laboratory, veterinary, crafts, Industrials, glue sealing. These caps are compatible for luer lock syringes and luer slip syringes both. Perfect for air-tight to prevent contents from drying up.
Catalog No. 50-252-2431 Supplier Med Vet International Supplier No. BUCWS100
These caps used for sealing syringes used in laboratory, veterinary, crafts, Industrials, glue sealing. These caps are compatible for luer lock syringes and luer slip syringes both. Perfect for air-tight to prevent contents from drying up.