Learn More
Waters Corp WATERS CORPORATION Tubing Assembly, Capillary, 25 μm x 30 in., BSM Connection
Catalog No. 50791932 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 25 μm x 30 in. capillary tubing assembly connects the Binary Solvent Manager (BSM) to the trap valve of the nanoACQUITY UPLC system. It is designed for specific system configurations to ensure proper fluidic connections.
  • Connects Binary Solvent Manager (BSM) to the trap valve.
  • Designed for nanoACQUITY UPLC system configurations.
  • Supports Dual Pump Trapping, Vented Dual Pump Trapping, and 2D configurations.
  • Features 25 μm diameter capillary tubing.
  • Measures 30 in. in length.
Catalog No. 50-791-932 Supplier Waters Corp Supplier No. 430001575
This 25 μm x 30 in. capillary tubing assembly connects the Binary Solvent Manager (BSM) to the trap valve of the nanoACQUITY UPLC system. It is designed for specific system configurations to ensure proper fluidic connections.
  • Connects Binary Solvent Manager (BSM) to the trap valve.
  • Designed for nanoACQUITY UPLC system configurations.
  • Supports Dual Pump Trapping, Vented Dual Pump Trapping, and 2D configurations.
  • Features 25 μm diameter capillary tubing.
  • Measures 30 in. in length.