Learn More
Waters Corp Waters Corporation Capillary Tubing, With Frit, 25μm × 18 in
Catalog No. NC9909732 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Capillary Tubing connects the BSM to the inject valve of the nanoACQUITY UPLC in the Direct Injection Configuration and the Single Pump Trapping Configuration. It also connects the Trap Valve with the MM union on the nanoACQUITY UPLC in the Dual Pump Trapping Configuration and the 2D Configuration.
  • Connects the BSM to the inject valve of the nanoACQUITY UPLC in direct injection configuration.
  • Connects the BSM to the inject valve of the nanoACQUITY UPLC in single pump trapping configuration.
  • Connects the trap valve with the MM union on the nanoACQUITY UPLC in dual pump trapping configuration.
  • Connects the trap valve with the MM union on the nanoACQUITY UPLC in 2D configuration.
  • Features 25μm × 18 in dimensions.
  • Supplied as 1 unit per package.
Catalog No. NC9909732 Supplier Waters Corp Supplier No. 430002242
This Capillary Tubing connects the BSM to the inject valve of the nanoACQUITY UPLC in the Direct Injection Configuration and the Single Pump Trapping Configuration. It also connects the Trap Valve with the MM union on the nanoACQUITY UPLC in the Dual Pump Trapping Configuration and the 2D Configuration.
  • Connects the BSM to the inject valve of the nanoACQUITY UPLC in direct injection configuration.
  • Connects the BSM to the inject valve of the nanoACQUITY UPLC in single pump trapping configuration.
  • Connects the trap valve with the MM union on the nanoACQUITY UPLC in dual pump trapping configuration.
  • Connects the trap valve with the MM union on the nanoACQUITY UPLC in 2D configuration.
  • Features 25μm × 18 in dimensions.
  • Supplied as 1 unit per package.