Learn More
Waters Corp Waters Corporation Tubing Assembly, Fused Silica, Inject Valve to Trap Valve, nanoACQUITY UPLC
Catalog No. 50791934 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed for use with the nanoACQUITY UPLC system in 2D analysis setups. It facilitates the connection between the injection valve and the trap valve, serving as an essential component within the online 2D-LC/MS Kit tubing assemblies.
  • Configures the nanoACQUITY UPLC system for 2D analysis.
  • Connects the injection valve to the trap valve.
  • Features fused silica tubing construction.
  • Equipped with a V-detail fitting at one end.
  • Assembled with a green PTFE sleeve and black plastic fitting into a SCX column.
  • V-detail end connects to port 4 of the HTM valve.
Catalog No. 50-791-934 Supplier Waters Corp Supplier No. 430001577
This tubing assembly is designed for use with the nanoACQUITY UPLC system in 2D analysis setups. It facilitates the connection between the injection valve and the trap valve, serving as an essential component within the online 2D-LC/MS Kit tubing assemblies.
  • Configures the nanoACQUITY UPLC system for 2D analysis.
  • Connects the injection valve to the trap valve.
  • Features fused silica tubing construction.
  • Equipped with a V-detail fitting at one end.
  • Assembled with a green PTFE sleeve and black plastic fitting into a SCX column.
  • V-detail end connects to port 4 of the HTM valve.