Learn More
Waters Corp Waters Corporation Tubing Assembly, Capillary, Injector to Trap Valve, 40 μm × 13 in.
Catalog No. 50791935 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This capillary tubing fits between the injector and the trap valve of the nanoACQUITY UPLC with dual pumps. It is used in the Vented Dual Pump and Dual Pump Trapping configurations. The tubing measures 40μm by 13 in. long.
  • Designed for nanoACQUITY UPLC systems with dual pumps
  • Ensures reliable connection between injector and trap valve
Catalog No. 50-791-935 Supplier Waters Corp Supplier No. 430001629
This capillary tubing fits between the injector and the trap valve of the nanoACQUITY UPLC with dual pumps. It is used in the Vented Dual Pump and Dual Pump Trapping configurations. The tubing measures 40μm by 13 in. long.
  • Designed for nanoACQUITY UPLC systems with dual pumps
  • Ensures reliable connection between injector and trap valve