Learn More
Thomas Scientific Cap, VersaCap 80, Open with Molded 1/2" HB, Adapter
SDP
Catalog No. 501458432
Encompass
Foxx Life Sciences’ VersaCaps and adapters allow users to easily customize carboys and Heavy Duty Vacuum Bottles for a variety of applications. VersaCap adapters are available with a variety of hose barb sizes and configurations, including molded in hose barbs and quick-connect hose barbs. VersaCaps are leakproof, and feature replaceable platinum-cured silicone gaskets, as well as large finger and hand grips for easy loosening and tightening. The Bioprocess adapter is compatible with custom closures that provide closed systems designed for fluid transfer. This is compatible with Saint-Gobain’s EZ TOP® Container Closures with C-FLEX® tubing. EZ TOP® and C-FLEX® are registered trademarks of Saint-Gobain. All VersaCaps and adapters are molded from autoclavable polypropylene, and are available in industry-standard 53B, 83B, and wide-mouth 120 mm sizes. The recommended autoclaving cycle is 20 minutes at 15 PSI and 121°C.
Catalog No. 50-145-8432 Supplier Thomas Scientific Supplier No. 1198J37
Foxx Life Sciences’ VersaCaps and adapters allow users to easily customize carboys and Heavy Duty Vacuum Bottles for a variety of applications. VersaCap adapters are available with a variety of hose barb sizes and configurations, including molded in hose barbs and quick-connect hose barbs. VersaCaps are leakproof, and feature replaceable platinum-cured silicone gaskets, as well as large finger and hand grips for easy loosening and tightening. The Bioprocess adapter is compatible with custom closures that provide closed systems designed for fluid transfer. This is compatible with Saint-Gobain’s EZ TOP® Container Closures with C-FLEX® tubing. EZ TOP® and C-FLEX® are registered trademarks of Saint-Gobain. All VersaCaps and adapters are molded from autoclavable polypropylene, and are available in industry-standard 53B, 83B, and wide-mouth 120 mm sizes. The recommended autoclaving cycle is 20 minutes at 15 PSI and 121°C.