Learn More
Chemglass Life Sciences Cap, Vented, 70mm, Blue, Polypropylene, Non-Sterile, 0.22µm PTFE Membrane
SDP
Catalog No. 50143769
Encompass_Preferred
Blue polypropylene vented cap has a 0. 22um PTFE hydrophobic membrane to allow gas exchange while preventing contamination.They are autoclavable at 121 degreesC.Must be used with70mm silicone gasket (listed below).
Catalog No. 50-143-769 Supplier Chemglass Life Sciences Supplier No. CLS1484A70
Blue polypropylene vented cap has a 0. 22um PTFE hydrophobic membrane to allow gas exchange while preventing contamination.They are autoclavable at 121 degreesC.Must be used with70mm silicone gasket (listed below).