Learn More
Hach Company Cap, white, pack of 6, for Culture Tube
Supplier:  Hach Company 2241106
Encompass
White cap for Culture Tube (product number 2275800 (pk/1)/ 2275806 (pk/6)). Screw cap has a firmly bonded fluoropolymer liner, providing a sure and highly inert sealing surface. Package of 6.
Catalog No. 50-199-5599