Learn More
Perkin Elmer US LLC 11 mm Pre-Assembled Blue Aluminum Crimp Cap with PTFE/Red Rubber Septa, 100/pk
Catalog No. 509052342
Encompass_Preferred
Our closures with septa are made in a wide variety of sizes and materials ensuring you have the right combination for your sample type and analysis temperature. Each product is manufactured using strict quality control standards resulting in consistent cap dimensions with septa which may be certified for higher purity grades. All our caps and septa are packed in a clean environment to ensure you receive contaminant free product every time. Vial Top Type Neck I.D. Size Cap Type Septa Material Quantity Crimp Top 11 mm Blue Aluminum PTFE/Red Rubber 100
Catalog No. 50-905-2342 Supplier Perkin Elmer US LLC Supplier No. N9302686
Our closures with septa are made in a wide variety of sizes and materials ensuring you have the right combination for your sample type and analysis temperature. Each product is manufactured using strict quality control standards resulting in consistent cap dimensions with septa which may be certified for higher purity grades. All our caps and septa are packed in a clean environment to ensure you receive contaminant free product every time. Vial Top Type Neck I.D. Size Cap Type Septa Material Quantity Crimp Top 11 mm Blue Aluminum PTFE/Red Rubber 100