Learn More
Aviva Systems Biology Corp CAP2 Antibody - C-terminal region (ARP63855_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63855_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CAP2. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-288-101