Learn More
Brookfield Engineer Labs CAP Mineral Oil Fluids, CAP3H
Catalog No. NC9952846
Encompass
AMETEK Brookfield Viscosity Standards are accurate to ±1% of the stated viscosity and are certified by methods traceable to the United States National Institute of Standards and Technology (NIST). The selection of one or two fluids will normally provide sufficient measurement points to verify the calibration of your instrument. CAP Oil Fluids are supplied in 150 mL (4 oz) containers.
Catalog No. NC9952846 Supplier Brookfield Engineer Labs Supplier No. CAP3H
AMETEK Brookfield Viscosity Standards are accurate to ±1% of the stated viscosity and are certified by methods traceable to the United States National Institute of Standards and Technology (NIST). The selection of one or two fluids will normally provide sufficient measurement points to verify the calibration of your instrument. CAP Oil Fluids are supplied in 150 mL (4 oz) containers.