Learn More
ATCC Capan-1; Pancreatic Carcinoma; Human (Homo sapiens)
Catalog No. 502382765
Encompass
Capan-1 is a cell line with epithelial morphology that was isolated from the pancreas of a 40-year-old, White, male with pancreatic adenocarcinoma. This cell line is a suitable transfection host.
Catalog No. 50-238-2765 Supplier ATCC Supplier No. HTB79
Capan-1 is a cell line with epithelial morphology that was isolated from the pancreas of a 40-year-old, White, male with pancreatic adenocarcinoma. This cell line is a suitable transfection host.