Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 100 x 4.6 mm

Catalog No. 502396290 Shop All Phenomenex Products
Encompass

Capcell Pak 3 m C18 MG-III 100 92766

Catalog No. 50-239-6290 Supplier Phenomenex Inc Supplier No. 92766
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Capcell Pak 3 m C18 MG-III 100 92766

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.