Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 100 x 1.5mm

Catalog No. 502396538 Shop All Phenomenex Products
Encompass

Capcell Pak 5u C18-UG 120A 70602

Catalog No. 50-239-6538 Supplier Phenomenex Inc Supplier No. 70602
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Capcell Pak 5u C18-UG 120A 70602

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.