Learn More
Bioworld Cape Sundew/ Venus Flytrap Pretransplant Basal Medium, 50 L
Catalog No. 50254886
Encompass_Preferred
Cape Sundew/ Venus Flytrap Pretransplant Basal MediumContains the Macronutrients, Micronutrients, Vitamins, & Plant Growth Regulators required for carnivorous plant growthSucrose, Cytokines, and Auxins should be added as neededProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 2.20 g/LpH: 4.5-5.5Soluble in water
Catalog No. 50-254-886 Supplier Bioworld Supplier No. 306301574
Cape Sundew/ Venus Flytrap Pretransplant Basal MediumContains the Macronutrients, Micronutrients, Vitamins, & Plant Growth Regulators required for carnivorous plant growthSucrose, Cytokines, and Auxins should be added as neededProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 2.20 g/LpH: 4.5-5.5Soluble in water