Learn More
Bioworld Cape Sundew/ Venus Flytrap Multiplication Basal Medium, 50 L
Catalog No. 50254883
Encompass_Preferred
Cape Sundew/ Venus Flytrap Multiplication Basal MediumContains the Macronutrients, Micronutrients, Vitamins, and Plant Growth Regulators required for carnivorous plant growthSucrose, Cytokines, and Auxins should be added as neededProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 2.12 g/LpH: 3.0-4.0Soluble in water
Catalog No. 50-254-883 Supplier Bioworld Supplier No. 306301564
Cape Sundew/ Venus Flytrap Multiplication Basal MediumContains the Macronutrients, Micronutrients, Vitamins, and Plant Growth Regulators required for carnivorous plant growthSucrose, Cytokines, and Auxins should be added as neededProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 2.12 g/LpH: 3.0-4.0Soluble in water