Learn More
Bulldog Bio Inc CAPEASY TOOL
SDP
Catalog No. NC1128858
Encompass_Preferred
The CapEasy was created to keep your daily labwork simple and less stressful. Sealing or removing the caps on 8-well and 12-well PCR strip tubes can often lead to loss of sample, cross contaminiation, and sore fingers. This can be avoided by using the CapEasy Tool. The uniformly distributed pressure delivered by this nifty product ensures perfect sealing of all wells regardless of domed-lids or flat-lids. When access to the contents of the strip tubes is needed, removing the lids is done in a single smooth motion without the chance of knocking the sample out of the tube.
Catalog No. NC1128858 Supplier Bulldog Bio Inc Supplier No. NGFGCDC01
The CapEasy was created to keep your daily labwork simple and less stressful. Sealing or removing the caps on 8-well and 12-well PCR strip tubes can often lead to loss of sample, cross contaminiation, and sore fingers. This can be avoided by using the CapEasy Tool. The uniformly distributed pressure delivered by this nifty product ensures perfect sealing of all wells regardless of domed-lids or flat-lids. When access to the contents of the strip tubes is needed, removing the lids is done in a single smooth motion without the chance of knocking the sample out of the tube.