Learn More
Selleck Chemical LLC Capecitabine S1156-10mM/1mL
SDP
Catalog No. 501360960
Encompass_Preferred
Capecitabine is a tumor-selective fluoropyrimidine carbamate which achieves higher intratumoral 5-FU level with lower toxicity than 5-FU Capecitabine treatment of HCT-15 cells causes condensation of DNA and induces apoptosis
Catalog No. 50-136-0960 Supplier Selleck Chemical LLC Supplier No. S115610MM/1ML
Capecitabine is a tumor-selective fluoropyrimidine carbamate which achieves higher intratumoral 5-FU level with lower toxicity than 5-FU Capecitabine treatment of HCT-15 cells causes condensation of DNA and induces apoptosis