Learn More
Medchemexpress LLC HY-B0016 5g , Capecitabine CAS:154361-50-9 Purity:98%
SDP
Supplier:  Medchemexpress LLC HYB00165G
Encompass_Preferred
Medchemexpress, HY-B0016 5g Capecitabine CAS:154361-50-9 Formula:C15H22FN3O6 DNA/RNA Synthesis Purity:98% Capecitabine is an oral prodrug that is converted to its only active metabolite, fluorouracil (FU), by thymidine phosphorylase. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2194