Learn More
Medchemexpress LLC Capecitabine-d11 | 1132662-08-8 | 99.9% | C15H11D11FN3O6 | 1 MG
SDP
Catalog No. 5000432553
Encompass_Preferred
Capecitabine-d11 is the deuterium labeled Capecitabine. Capecitabine is an oral proagent that is converted to its active metabolite, 5-FU, by thymidine phosphorylase. It is for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-32553 Supplier Medchemexpress LLC Supplier No. HYB0016S1MG
Capecitabine-d11 is the deuterium labeled Capecitabine. Capecitabine is an oral proagent that is converted to its active metabolite, 5-FU, by thymidine phosphorylase. It is for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.