Learn More
Cayman Chemical Capecitabine, Purity: ≥98%, CAS: 154361-50-9, 5 g
SDP
Catalog No. 502890044
Encompass_Preferred
A prodrug of 5-fluorouracil; cytotoxic only at high concentrations in Scaber, SIHA, and MKN45 cells (IC50s = 97, 578, and 994 µM, respectively); inactive in a variety of cancer cell lines, including COLO205, HCT116, and MCF-7 cells (IC50s = >1,000 µM)
Catalog No. 50-289-0044 Supplier Cayman Chemical Supplier No. 104875 G
A prodrug of 5-fluorouracil; cytotoxic only at high concentrations in Scaber, SIHA, and MKN45 cells (IC50s = 97, 578, and 994 µM, respectively); inactive in a variety of cancer cell lines, including COLO205, HCT116, and MCF-7 cells (IC50s = >1,000 µM)