Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Capecitabine (Standard) | 154361-50-9 | 99.99% | C15H22FN3O6 | 100 MG
SDP

Catalog No. 5000433417
Encompass_Preferred

Capecitabine (Standard) is the analytical standard of Capecitabine, an oral proagent. It is designed for research and analytical applications.

  • Capecitabine is converted to its active metabolite, 5-FU, by thymidine phosphorylase.
  • It is commonly used in qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS.
  • The product appears as a solid, with a color ranging from white to off-white.
  • Molecular Weight: 359.35.
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: Please store the product under the recommended conditions specified in the Certificate of Analysis (COA).

Catalog No. 50-004-33417 Supplier Medchemexpress LLC Supplier No. HYB0016R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Capecitabine (Standard) is the analytical standard of Capecitabine, an oral proagent. It is designed for research and analytical applications.

  • Capecitabine is converted to its active metabolite, 5-FU, by thymidine phosphorylase.
  • It is commonly used in qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS.
  • The product appears as a solid, with a color ranging from white to off-white.
  • Molecular Weight: 359.35.
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: Please store the product under the recommended conditions specified in the Certificate of Analysis (COA).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.