Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capecitabine, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18601414 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-414 500 g Amber Glass Bottle
18-601-415 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-601-414 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C2417500GMBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

C2417, 154361-50-9, C15H22FN3O6

Spectrum™ Chemical Capecitabine, USP is used to treat breast, colon and rectal cancer cells by slowing or stopping their growth. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 154361-50-9
Molecular Formula C15H22FN3O6
Molecular Weight (g/mol) 359.35
InChI Key GAGWJHPBXLXJQN-UORFTKCHSA-N
IUPAC Name pentyl N-{1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-2-oxo-1,2-dihydropyrimidin-4-yl}carbamate
SMILES CCCCCOC(=O)NC1=NC(=O)N(C=C1F)[C@@H]1O[C@H](C)[C@@H](O)[C@H]1O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 98 to 102%
Grade USP
Packaging Amber Glass Bottle
Specific Rotation +96.0° to +100.0°
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.