Learn More
Cayman Chemical Capecitabined11 GC MS Standard
SDP
Supplier:  Cayman Chemical 100106811 MG
Encompass_Preferred
An internal standard for the quantification of capecitabine by GC- or LC-MS
Catalog No. 50-289-0288