Learn More
SAI Infusion Technologies 300uL Serum Blood Tube
SDP
Catalog No. NC1837148
Encompass
300uL Blood Collection Tube with Clotting Activator for Serum sampling. For collection of blood in preclinical studies via drip, transfer, or capillary action. Supplied with a carrier tube for transport and centrifugation. Optimized tube geometry ensures maximum plasma recovery. 100/box. Product is for laboratory animal research only.
This item is not returnable. View return policy
Catalog No. NC1837148 Supplier SAI Infusion Technologies Supplier No. MVCBS300
300uL Blood Collection Tube with Clotting Activator for Serum sampling. For collection of blood in preclinical studies via drip, transfer, or capillary action. Supplied with a carrier tube for transport and centrifugation. Optimized tube geometry ensures maximum plasma recovery. 100/box. Product is for laboratory animal research only.