Learn More
BUCHI Capillaries for Melting Point Apparatus
GSA_VASDP
Catalog No. 05402779 Shop All BUCHI Products
Change view
Click to view available options
Packaging Quantity:
100 Tubes
1000 Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
05-402-779 100 Tubes
05-402-710 1000 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-402-779 Supplier BUCHI Supplier No. 017808
Add to Cart
Add to Cart
For use with BUCHI Melting Point apparatus.
  • High-quality Borosilicate 3.3 glass
  • Optimized thickness
  • Reliable results
  • Compatible with other automated instrument or manual techniques
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Melting Point Capillary
Dimensions (L x W x H) 180 x 120 x 80 mm
For Use With (Equipment) BUCHI Melting Point Apparatus
Height (Metric) 80 mm
Material Glass
Packaging Quantity 100 Tubes