Learn More
Agilent Technologies CAPILLARY, 0.17MM, 500MM
Catalog No. NC1814499
Encompass
Capillary, stainless steel, 0.17mm id, 500 mm length, with 1 x stainless steel long screw fitting for 10-32 thread size, and 1 x M4 thread size fitting
Catalog No. NC1814499 Supplier Agilent Technologies Supplier No. 50676188
Capillary, stainless steel, 0.17mm id, 500 mm length, with 1 x stainless steel long screw fitting for 10-32 thread size, and 1 x M4 thread size fitting