Learn More
Molex Molex-Polymicro Capillary 100UMx193UM 25M Reel
Catalog No. 501108604
Encompass_Preferred
TSP100200 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 100um ID x 193um OD, spool length of 25m.
Catalog No. 50-110-8604 Supplier Molex Supplier No. 1068150021/25M
TSP100200 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 100um ID x 193um OD, spool length of 25m.