Learn More
Molex Molex-Polymicro Capillary 100UMx193UM 50M Reel
Catalog No. 501107742
Encompass_Preferred
TSP100200 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 100um ID x 193um OD, spool length of 50m.
Catalog No. 50-110-7742 Supplier Molex Supplier No. 1068150021/50M
TSP100200 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 100um ID x 193um OD, spool length of 50m.