Learn More
Molex Molex-Polymicro Capillary 100UMx363UM 25M Reel
Catalog No. 501108605
Encompass_Preferred
TSP100375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 100um ID x 363um OD, spool length of 25m.
Catalog No. 50-110-8605 Supplier Molex Supplier No. 1068150023/25M
TSP100375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 100um ID x 363um OD, spool length of 25m.