Learn More
Molex Molex-Polymicro Capillary 100UMx363UM 50M Reel
Catalog No. 501108623
Encompass_Preferred
TSP100375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 100um ID x 363um OD, spool length of 50m.
Catalog No. 50-110-8623 Supplier Molex Supplier No. 1068150023/50M
TSP100375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 100um ID x 363um OD, spool length of 50m.