Learn More
Molex Molex-Polymicro Capillary 10UMx150UM 25M Reel
Catalog No. 501107694
Encompass_Preferred
TSP010150 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 10um ID x 150um OD, spool length of 25m.
Catalog No. 50-110-7694 Supplier Molex Supplier No. 1068150004/25M
TSP010150 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 10um ID x 150um OD, spool length of 25m.