Learn More
Molex Molex-Polymicro Capillary 10UMx363UM 25M Reel
Catalog No. 501108589
Encompass_Preferred
TSP010375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 10um ID x 363um OD, spool length of 25m.
Catalog No. 50-110-8589 Supplier Molex Supplier No. 1068150005/25M
TSP010375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 10um ID x 363um OD, spool length of 25m.