Learn More
Molex Molex-Polymicro Capillary 10UMx363UM 50M Reel
Catalog No. 501107724
Encompass_Preferred
TSP010375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 10um ID x 363um OD, spool length of 50m..
Catalog No. 50-110-7724 Supplier Molex Supplier No. 1068150005/50M
TSP010375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 10um ID x 363um OD, spool length of 50m..