Learn More
Molex Molex-Polymicro Capillary 150UMx363UM 25M Reel
Catalog No. 501108606
Encompass_Preferred
TSP150375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 150um ID x 363um OD, spool length of 25m.
Catalog No. 50-110-8606 Supplier Molex Supplier No. 1068150024/25M
TSP150375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 150um ID x 363um OD, spool length of 25m.