Learn More
Molex Molex-Polymicro Capillary 15UMx363UM 25M Reel
Catalog No. 501108590
Encompass_Preferred
TSP015375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 15um ID x 363um OD, spool length of 25m.
Catalog No. 50-110-8590 Supplier Molex Supplier No. 1068150007/25M
TSP015375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 15um ID x 363um OD, spool length of 25m.