Learn More
Molex Molex-Polymicro Capillary 180UMx360UM 50M Reel
Catalog No. 501108624
Encompass_Preferred
TSP180350 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 180um ID x 360um OD, spool length of 50m.
Catalog No. 50-110-8624 Supplier Molex Supplier No. 1068150025/50M
TSP180350 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 180um ID x 360um OD, spool length of 50m.