Learn More
Molex Molex-Polymicro Capillary 200UMx360UM 50M Reel
Supplier:  Molex 1068150204/50M
Encompass_Preferred
TSP200350 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 200um ID x 360um OD, spool length of 50m.
Catalog No. 50-110-7746