Learn More
Molex Molex-Polymicro Capillary 20UMx363UM 50M Reel
Catalog No. 501108615
Encompass_Preferred
TSP020375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 20um ID x 363um OD, spool length of 50m.
Catalog No. 50-110-8615 Supplier Molex Supplier No. 1068150009/50M
TSP020375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 20um ID x 363um OD, spool length of 50m.