Learn More
Molex Molex-Polymicro Capillary 20UMx90UM 25M Reel
Catalog No. 501107698
Encompass_Preferred
TSP020090 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 20um ID x 90um OD, spool length of 25m.
Catalog No. 50-110-7698 Supplier Molex Supplier No. 1068150381/25M
TSP020090 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 20um ID x 90um OD, spool length of 25m.