Learn More
Molex Molex-Polymicro Capillary 20UMx90UM 50M Reel
Catalog No. 501108614
Encompass_Preferred
TSP020090 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 20um ID x 90um OD, spool length of 50m.
Catalog No. 50-110-8614 Supplier Molex Supplier No. 1068150381/50M
TSP020090 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 20um ID x 90um OD, spool length of 50m.