Learn More
Molex Molex-Polymicro Capillary 250UMx360UM 25M Reel
Catalog No. 501108609
Encompass_Preferred
TSP250350 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 250um ID x 360um OD, spool length of 25m.
Catalog No. 50-110-8609 Supplier Molex Supplier No. 1068150026/25M
TSP250350 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 250um ID x 360um OD, spool length of 25m.