Learn More
Molex Molex-Polymicro Capillary 250UMx360UM 10M Reel
Supplier:  Molex 1068150026/10M
Encompass_Preferred
TSP250350 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 250um ID x 360um OD, spool length of 10m.
Catalog No. 50-110-8586