Learn More
Molex Molex-Polymicro Capillary 25UMx150UM 10M Reel
Supplier:  Molex 1068151145/10M
Encompass_Preferred
TSP025150 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 25um ID x 150um OD, spool length of 10m.
Catalog No. 50-110-8577