Learn More
Molex Molex-Polymicro Capillary 25UMx363UM 10M Reel
Supplier:  Molex 1068150011/10M
Encompass_Preferred
TSP025375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 25um ID x 363um OD, spool length of 10m.
Catalog No. 50-110-8578