Learn More
Molex Molex-Polymicro Capillary 25UMx363UM 25M Reel
Catalog No. 501108593
Encompass_Preferred
TSP025375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 25um ID x 363um OD, spool length of 25m.
Catalog No. 50-110-8593 Supplier Molex Supplier No. 1068150011/25M
TSP025375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 25um ID x 363um OD, spool length of 25m.