Learn More
Molex Molex-Polymicro Capillary 30UMx150UM 25M Reel
Catalog No. 501108594
Encompass_Preferred
TSP030150 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 30um ID x 150um OD, spool length of 25m.
Catalog No. 50-110-8594 Supplier Molex Supplier No. 1068150012/25M
TSP030150 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 30um ID x 150um OD, spool length of 25m.