Learn More
Molex Molex-Polymicro Capillary 320UMx435UM 25M Reel
Catalog No. 501108610
Encompass_Preferred
TSP320450 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 320um ID x 435um OD, spool length of 25m.
Catalog No. 50-110-8610 Supplier Molex Supplier No. 1068150027/25M
TSP320450 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 320um ID x 435um OD, spool length of 25m.