Learn More
Molex Molex-Polymicro Capillary 320UMx435UM 50M Reel
Catalog No. 501108625
Encompass_Preferred
TSP320450 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 320um ID x 435um OD, spool length of 50m.
Catalog No. 50-110-8625 Supplier Molex Supplier No. 1068150027/50M
TSP320450 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 320um ID x 435um OD, spool length of 50m.